4-Chlor-2,6-xylenol 98%

Lieferant: Apollo Scientific
Achtung

OR322178-1G OR322178-5G OR322178-25G
APOSOR322178-1GEA 21.9 EUR
APOSOR322178-1G APOSOR322178-5G APOSOR322178-25G
4-Chlor-2,6-xylenol 98%
4-Chlor-2,6-xylenol
Formel: C₈H₉ClO
Molecular Weight: 156.61 g/mol
Schmelzpunkt: 81…83 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00020138
CAS-Nummer: 1123-63-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR