2-Ethoxy-4-methylphenol

Lieferant: Apollo Scientific

OR322189-1G OR322189-10G
APOSOR322189-1GEA 58 EUR
APOSOR322189-1G APOSOR322189-10G
2-Ethoxy-4-methylphenol
2-Ethoxy-4-methylphenol
Formel: C₉H₁₂O₂
Molecular Weight: 152.19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00043697
CAS-Nummer: 2563-07-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR