1-Adamantyl isocyanate

Lieferant: Apollo Scientific
Achtung

Synonyme: Isocyanic acid 1-Adamantyl Ester, 1-Adamantyl Isocyanate

OR322196-250MG OR322196-5G
APOSOR322196-250MGEA 58.2 EUR
APOSOR322196-250MG APOSOR322196-5G
1-Adamantyl isocyanate
1-Adamantyl isocyanate

When melted, may contain small amount of sediment.

Formel: C₁₁H₁₅NO
Molecular Weight: 177,247 g/mol
Schmelzpunkt: 147 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00134405
CAS-Nummer: 4411-25-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR