Monomethylphthalat

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Methoxycarbonyl)benzoic acid, Methyl hydrogen phthalate

OR322213-100G OR322213-500G
APOSOR322213-100GEA 149 EUR
APOSOR322213-100G APOSOR322213-500G
Monomethylphthalat
Monomethylphthalat
Formel: C₉H₈O₄
Molecular Weight: 180,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00002466
CAS-Nummer: 4376-18-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR