(S)-Methyl-2-amino-3-phenylpropanoate hydrochloride 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: L-Methyl phenylalanate hydrochloride, H-Phe-OMe·HCl

OR322217-500G
APOSOR322217-500GEA 72.5 EUR
APOSOR322217-500G
(S)-Methyl-2-amino-3-phenylpropanoate hydrochloride 98%
(S)-Methyl-2-amino-3-phenylpropanoate hydrochloride
Formel: C₁₀H₁₄ClNO₂
Molecular Weight: 215,68 g/mol
Schmelzpunkt: 158…162 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012489
CAS-Nummer: 7524-50-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR