N-(Benzyloxycarbonyloxy)succinimid 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
CBZ-OSu (Z-OSu)
OR322228-100G
OR322228-25G
APOSOR322228-100GEA
24.8
EUR
APOSOR322228-100G
APOSOR322228-25G
N-(Benzyloxycarbonyloxy)succinimid 97%
N-(Benzyloxycarbonyloxy)succinimid
Formel:
C₁₂H₁₁NO₅ Molecular Weight: 249,22 g/mol Schmelzpunkt: 80…82 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005513 CAS-Nummer: 13139-17-8 |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...