Ethyl-2-isocyanatobenzoat 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Ethoxycarbonyl)phenyl isocyanate

OR322234-1G OR322234-25G OR322234-5G
APOSOR322234-1GEA 52.2 EUR
APOSOR322234-1G APOSOR322234-25G APOSOR322234-5G
Ethyl-2-isocyanatobenzoat 97%
Ethyl-2-isocyanatobenzoat

When melted, may contain small amount of sediment.

Formel: C₁₀H₉NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00013849
CAS-Nummer: 76393-16-3
UN: 3335
ADR: 9,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR