3-(Dimethylamino)benzoylchlorid Hydrochlorid 90%
Lieferant: Apollo Scientific
|
Gefahr
|
OR322248-1G
OR322248-5G
APOSOR322248-1GEA
0
EUR
APOSOR322248-1G
APOSOR322248-5G
3-(Dimethylamino)benzoylchlorid Hydrochlorid 90%
3-(Dimethylamino)benzoylchlorid Hydrochlorid
Formel:
C₉H₁₁Cl₂NO Molecular Weight: 220,1 g/mol Schmelzpunkt: 131…133 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD19442144 CAS-Nummer: 117500-61-5 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...