Methyl-2,2-dimethylacetoacetate

Lieferant: Apollo Scientific

Synonyme: Methyl 2,2-dimethylacetoacetate

OR322264-1G
APOSOR322264-1GEA 63.8 EUR
APOSOR322264-1G
Methyl-2,2-dimethylacetoacetate
Methyl-2,2-dimethylacetoacetate
Formel: C₇H₁₂O₃
Molecular Weight: 144.17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00026232
CAS-Nummer: 38923-57-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR