2-Hydroxy-5-methylisophthalaldehyd 95%

Lieferant: Apollo Scientific

Synonyme: 2-Hydroxy-5-methylbenzene-1,3-dicarbaldehyde

OR32232-5G
APOSOR32232-5GEA 140 EUR
APOSOR32232-5G
2-Hydroxy-5-methylisophthalaldehyd 95%
2-Hydroxy-5-methylisophthalaldehyd
Formel: C₉H₈O₃
Molecular Weight: 164,1600 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00016596
CAS-Nummer: 7310-95-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR