5-Bromothiophene-2-carboxaldoxime
Lieferant: Apollo Scientific
Synonyme:
5-Bromothiophene-2-carbaldehyde oxime
OR322321-500MG
OR322321-1G
APOSOR322321-500MGEA
50.9
EUR
APOSOR322321-500MG
APOSOR322321-1G
5-Bromothiophene-2-carboxaldoxime
5-Bromothiophene-2-carboxaldoxime
Formel:
C₅H₄BrNOS Molecular Weight: 206.06 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00126375 CAS-Nummer: 2160-63-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...