4-(Morpholinomethyl)aniline
Lieferant: Apollo Scientific
|
Achtung
|
OR322351-1G
OR322351-5G
APOSOR322351-1GEA
21.9
EUR
APOSOR322351-1G
APOSOR322351-5G
4-(Morpholinomethyl)aniline
4-(Morpholinomethyl)aniline
Formel:
C₁₁H₁₆N₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD03659020 CAS-Nummer: 51013-67-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...