1-(4-Nitrophenyl)-1H-pyrrol 95%
Lieferant: Apollo Scientific
Synonyme:
1-(4-Nitrophenyl)pyrrole
OR32237-25G
APOSOR32237-25GEA
126
EUR
APOSOR32237-25G
1-(4-Nitrophenyl)-1H-pyrrol 95%
1-(4-Nitrophenyl)-1H-pyrrole
Formel:
C₁₀H₈N₂O₂ Molecular Weight: 188,1800 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00119340 CAS-Nummer: 4533-42-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...