3-Pyrid-2-ylbenzoesäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(Pyridin-2-yl)benzoic acid

OR322374-1G OR322374-5G
APOSOR322374-1GEA 52.2 EUR
APOSOR322374-1G APOSOR322374-5G
3-Pyrid-2-ylbenzoesäure 98%
3-Pyrid-2-ylbenzoesäure
Formel: C₁₂H₉NO₂
Molecular Weight: 199,2054 g/mol
Schmelzpunkt: 207 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05864807
CAS-Nummer: 4467-07-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR