2,5-Dimethylresorcin
Lieferant: Apollo Scientific
Synonyme:
2,5-Dimethylbenzene-1,3-diol
OR322380-1G
OR322380-250MG
OR322380-5G
APOSOR322380-1GEA
679
EUR
APOSOR322380-1G
APOSOR322380-250MG
APOSOR322380-5G
2,5-Dimethylresorcin
2,5-Dimethylresorcin
Formel:
C₈H₁₀O₂ Molecular Weight: 138,1638 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00134473 CAS-Nummer: 488-87-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...