2-[(6-chloropyridazin-3-yl)amino]ethanol 95%

Lieferant: Apollo Scientific

OR32239-1G OR32239-5G
APOSOR32239-1GEA 224 EUR
APOSOR32239-1G APOSOR32239-5G
2-[(6-chloropyridazin-3-yl)amino]ethanol 95%
2-[(6-chloropyridazin-3-yl)amino]ethanol
Formel: C₆H₈ClN₃O
Molecular Weight: 173,6 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 51947-89-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR