4-Methyl-2-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)pentanoic acid 95%

Lieferant: Apollo Scientific

OR32241-1G OR32241-5G
APOSOR32241-1GEA 180 EUR
APOSOR32241-1G APOSOR32241-5G
4-Methyl-2-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)pentanoic acid 95%
4-Methyl-2-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)pentanoic acid
Formel: C₁₄H₁₇NO₃
Molecular Weight: 247.3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02082683
CAS-Nummer: 298700-70-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR