3-(Piperidin-4-yl)benzonitrile

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(3-Cyanophenyl)piperidine

OR322434-1G OR322434-250MG
APOSOR322434-1GEA 661 EUR
APOSOR322434-1G APOSOR322434-250MG
3-(Piperidin-4-yl)benzonitrile
3-(Piperidin-4-yl)benzonitrile
Formel: C₁₂H₁₄N₂
Molecular Weight: 186,26 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD03839908
CAS-Nummer: 370864-72-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR