3-Nitrosalicylaldehyd 99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Hydroxy-3-nitrobenzaldehyde
OR322457-1G
OR322457-5G
APOSOR322457-1GEA
53.8
EUR
APOSOR322457-1G
APOSOR322457-5G
3-Nitrosalicylaldehyd 99%
3-Nitrosalicylaldehyd
Formel:
C₇H₅NO₄ Molecular Weight: 167,12 g/mol Schmelzpunkt: 105…110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041874 CAS-Nummer: 5274-70-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...