2-(2-Propynyloxy)benzenecarboxylic acid 95%

Lieferant: Apollo Scientific

OR32246-1G OR32246-5G
APOSOR32246-1GEA 336 EUR
APOSOR32246-1G APOSOR32246-5G
2-(2-Propynyloxy)benzenecarboxylic acid 95%
2-(2-Propynyloxy)benzenecarboxylic acid
Formel: C₁₀H₈O₃
Molecular Weight: 176.17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03001228
CAS-Nummer: 55582-31-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR