1-[4-(1H-Pyrrol-1-yl)phenyl]-1-ethanone 95%

Lieferant: Apollo Scientific

Synonyme: 1-(4-(1H-Pyrrol-1-yl)phenyl)ethanone

OR32249-1G OR32249-5G
APOSOR32249-1GEA 270 EUR
APOSOR32249-1G APOSOR32249-5G
1-[4-(1H-Pyrrol-1-yl)phenyl]-1-ethanone 95%
1-[4-(1H-Pyrrol-1-yl)phenyl]-1-ethanone
Formel: C₁₂H₁₁NO
Molecular Weight: 185.23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02572018
CAS-Nummer: 22106-37-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR