1-Hydroxy-6-methylsulphonylindole 95+%

Lieferant: Apollo Scientific

OR3225-25G OR3225-5G OR3225-1G
APOSOR3225-25GEA 734 EUR
APOSOR3225-25G APOSOR3225-5G APOSOR3225-1G
1-Hydroxy-6-methylsulphonylindole 95+%
1-Hydroxy-6-methylsulphonylindole
Formel: C₉H₉NO₃S
Molecular Weight: 211,24 g/mol
Schmelzpunkt: 174…176 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00216482
CAS-Nummer: 170492-47-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR