[4-Methoxy-3-(2-propynyloxy)phenyl]methanol 95%

Lieferant: Apollo Scientific

OR32250-1G OR32250-5G
APOSOR32250-1GEA 336 EUR
APOSOR32250-1G APOSOR32250-5G
[4-Methoxy-3-(2-propynyloxy)phenyl]methanol 95%
[4-Methoxy-3-(2-propynyloxy)phenyl]methanol
Formel: C₁₁H₁₂O₃
Molecular Weight: 192.22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03001238
CAS-Nummer: 385383-49-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR