4-Methoxy-3-(2-propynyloxy)benzenecarbaldehydeoxime 95%

Lieferant: Apollo Scientific

Synonyme: (E)-N-{[4-Methoxy-3-(prop-2-yn-1-yloxy)phenyl]methylidene}hydroxylamine

OR32251-1G OR32251-5G
APOSOR32251-1GEA 336 EUR
APOSOR32251-1G APOSOR32251-5G
4-Methoxy-3-(2-propynyloxy)benzenecarbaldehydeoxime 95%
4-Methoxy-3-(2-propynyloxy)benzenecarbaldehydeoxime
Formel: C₁₁H₁₁NO₃
Molecular Weight: 205,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03001240
CAS-Nummer: 385383-47-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR