(3R,4R)-1-Benzylpyrrolidine-3,4-diol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(3R,4R)-(-)-1-Benzyl-3,4-pyrrolidindiol
OR322563-5G
APOSOR322563-5GEA
206
EUR
APOSOR322563-5G
(3R,4R)-1-Benzylpyrrolidine-3,4-diol
(3R,4R)-1-Benzylpyrrolidine-3,4-diol
Formel:
C₁₁H₁₅NO₂ Molecular Weight: 193,2423 g/mol Schmelzpunkt: 94…100 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01073894 CAS-Nummer: 163439-82-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...