2-Bromo-1-(4-methoxy-3-nitrophenyl)-1-ethanone
Lieferant: Apollo Scientific
|
Gefahr
|
OR322579-250MG
OR322579-100MG
OR322579-1G
APOSOR322579-250MGEA
308
EUR
APOSOR322579-250MG
APOSOR322579-100MG
APOSOR322579-1G
2-Bromo-1-(4-methoxy-3-nitrophenyl)-1-ethanone
2-Bromo-1-(4-methoxy-3-nitrophenyl)-1-ethanone
Formel:
C₉H₈BrNO₄ Molecular Weight: 274,08 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06666938 CAS-Nummer: 65447-49-6 UN: 1759 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...