1-Ethyl-3-methyl-1H-pyrazol-5-amine 95%

Lieferant: Apollo Scientific

OR32269-1G OR32269-5G
APOSOR32269-1GEA 270 EUR
APOSOR32269-1G APOSOR32269-5G
1-Ethyl-3-methyl-1H-pyrazol-5-amine 95%
1-Ethyl-3-methyl-1H-pyrazol-5-amine
Formel: C₆H₁₁N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD02255831
CAS-Nummer: 3524-33-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR