4-(4-Methylpiperazino)-1,2-benzenediamine 95%
Lieferant: Apollo Scientific
OR322770-1G
OR322770-250MG
APOSOR322770-1GEA
434
EUR
APOSOR322770-1G
APOSOR322770-250MG
4-(4-Methylpiperazino)-1,2-benzenediamine 95%
4-(4-Methylpiperazino)-1,2-benzenediamine
Formel:
C₁₁H₁₈N₄ Schmelzpunkt: 101…103 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03001209 CAS-Nummer: 54998-08-2 UN: 3259 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...