4-Chloro-6-(4-methyl-1-piperazinyl)pyrimidine 96%

Lieferant: Apollo Scientific

OR322861-1G OR322861-5G
APOSOR322861-1GEA 201 EUR
APOSOR322861-1G APOSOR322861-5G
4-Chloro-6-(4-methyl-1-piperazinyl)pyrimidine 96%
4-Chloro-6-(4-methyl-1-piperazinyl)pyrimidine
Formel: C₉H₁₃ClN₄
Molecular Weight: 212.68 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05739691
CAS-Nummer: 27174-60-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR