DL-Isoserin 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
H-DL-IsoSer-OH, DL-3-Amino-2-hydroxypropionic Acid
OR322969-10G
OR322969-5G
APOSOR322969-10GEA
68.3
EUR
APOSOR322969-10G
APOSOR322969-5G
DL-Isoserin 97%
DL-Isoserine
Formel:
C₃H₇NO₃ Molecular Weight: 105,09 g/mol Schmelzpunkt: 235 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008138 CAS-Nummer: 565-71-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...