5-Amino-2-(4-aminophenyl)benzimidazole 98+%

Lieferant: Apollo Scientific

Synonyme: 2-(4-Aminophenyl)-1H-benzimidazol-5-amine

OR323128-25G
APOSOR323128-25GEA 62.5 EUR
APOSOR323128-25G
5-Amino-2-(4-aminophenyl)benzimidazole 98+%
5-Amino-2-(4-aminophenyl)benzimidazole
Formel: C₁₃H₁₂N₄
Molecular Weight: 224,27 g/mol
Schmelzpunkt: 236 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00451475
CAS-Nummer: 7621-86-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR