1-(2-Propynyl)-1H-indole-2,3-dione 95%

Lieferant: Apollo Scientific

Synonyme: 1-(Prop-2-yn-1-yl)-2,3-dihydro-1H-indole-2,3-dione

OR32315-1G OR32315-5G
APOSOR32315-1GEA 180 EUR
APOSOR32315-1G APOSOR32315-5G
1-(2-Propynyl)-1H-indole-2,3-dione 95%
1-(2-Propynyl)-1H-indole-2,3-dione
Formel: C₁₁H₇NO₂
Molecular Weight: 185.18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00225166
CAS-Nummer: 4290-87-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR