1-Chloro-3,4-dihydro-2-naphthalenecarbaldehyde 95%
Lieferant: Apollo Scientific
Synonyme:
1-Chloro-3,4-dihydronaphthalene-2-carbaldehyde
OR32322-1G
OR32322-5G
APOSOR32322-1GEA
336
EUR
APOSOR32322-1G
APOSOR32322-5G
1-Chloro-3,4-dihydro-2-naphthalenecarbaldehyde 95%
1-Chloro-3,4-dihydro-2-naphthalenecarbaldehyde
Formel:
C₁₁H₉ClO Molecular Weight: 192,65 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02257735 CAS-Nummer: 3262-03-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...