8-Anilino-1-naphthalinsulfonsäure 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 8-Anilinonaphthalene-1-sulphonic acid, 8-(Phenylamino)naphthalene-1-sulphonic acid

OR323280-100G OR323280-25G OR323280-5G
APOSOR323280-100GEA 166 EUR
APOSOR323280-100G APOSOR323280-25G APOSOR323280-5G
8-Anilino-1-naphthalinsulfonsäure 95%
8-Anilino-1-naphthalinsulfonsäure
Formel: C₁₆H₁₃NO₃S
Schmelzpunkt: 215…217 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003998
CAS-Nummer: 82-76-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR