1-[(6-Chloropyridin-3-yl)methyl]-4-methylpiperazine 95%

Lieferant: Apollo Scientific

OR32353-5G
APOSOR32353-5GEA 270 EUR
APOSOR32353-5G
1-[(6-Chloropyridin-3-yl)methyl]-4-methylpiperazine 95%
1-[(6-Chloropyridin-3-yl)methyl]-4-methylpiperazine
Formel: C₁₁H₁₆ClN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD04124601
CAS-Nummer: 612487-31-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR