4-[(6-Chloropyridin-3-yl)methyl]morpholine 95%
Lieferant: Apollo Scientific
Synonyme:
4-((6-Chloropyridin-3-yl)methyl)morpholine
OR32354-1G
OR32354-5G
APOSOR32354-1GEA
180
EUR
APOSOR32354-1G
APOSOR32354-5G
4-[(6-Chloropyridin-3-yl)methyl]morpholine 95%
4-[(6-Chloropyridin-3-yl)methyl]morpholine
Formel:
C₁₀H₁₃ClN₂O Molecular Weight: 212,68 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00684829 CAS-Nummer: 311774-34-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...