3-(Dimethylamino)-2-phenylacrylonitrile 95%

Lieferant: Apollo Scientific

Synonyme: (2Z)-3-(Dimethylamino)-2-phenylprop-2-enenitrile

OR32373-1G OR32373-25G OR32373-5G
APOSOR32373-1GEA 180 EUR
APOSOR32373-1G APOSOR32373-25G APOSOR32373-5G
3-(Dimethylamino)-2-phenylacrylonitrile 95%
3-(Dimethylamino)-2-phenylacrylonitrile
Formel: C₁₁H₁₂N₂
Molecular Weight: 172,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01935969
CAS-Nummer: 18226-50-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR