1-[(Ammoniooxy)methyl]-2-methoxybenzolchlorid 95%

Lieferant: Apollo Scientific

Synonyme: O-[(2-Methoxyphenyl)methyl]hydroxylamine hydrochloride

OR32399-1G OR32399-5G
APOSOR32399-1GEA 336 EUR
APOSOR32399-1G APOSOR32399-5G
1-[(Ammoniooxy)methyl]-2-methoxybenzolchlorid 95%
1-[(Ammoniooxy)methyl]-2-methoxybenzolchlorid
Formel: C₈H₁₂ClNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01114579
CAS-Nummer: 317821-72-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR