1-Methyl-3-phenyl-1H-pyrazol-5-ol 95%

Lieferant: Apollo Scientific

OR32409-1G OR32409-25G OR32409-5G
APOSOR32409-1GEA 180 EUR
APOSOR32409-1G APOSOR32409-25G APOSOR32409-5G
1-Methyl-3-phenyl-1H-pyrazol-5-ol 95%
1-Methyl-3-phenyl-1H-pyrazol-5-ol
Formel: C₁₀H₁₀N₂O
Molecular Weight: 174.2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03001245
CAS-Nummer: 34347-81-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR