Ethyl-2-[(2-chlorobenzoyl)amino]acetate 95%
Lieferant: Apollo Scientific
Synonyme:
Ethyl 2-[(2-chlorophenyl)formamido]acetate
OR32422-1G
OR32422-25G
OR32422-5G
APOSOR32422-1GEA
180
EUR
APOSOR32422-1G
APOSOR32422-25G
APOSOR32422-5G
Ethyl-2-[(2-chlorobenzoyl)amino]acetate 95%
Ethyl-2-[(2-chlorobenzoyl)amino]acetate
Formel:
C₁₁H₁₂ClNO₃ Molecular Weight: 241,68 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00793745 CAS-Nummer: 66824-94-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...