2-Methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarbonitrile 95%
Lieferant: Apollo Scientific
OR32433-1G
OR32433-5G
APOSOR32433-1GEA
336
EUR
APOSOR32433-1G
APOSOR32433-5G
2-Methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarbonitrile 95%
2-Methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarbonitrile
Formel:
C₇H₈N₂O Molecular Weight: 136.15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00974996 CAS-Nummer: 27036-90-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...