2-(2,5-Dimethyl-1H-pyrrol-1-yl)phenylamine 95%
Lieferant: Apollo Scientific
Synonyme:
2-(2,5-Dimethyl-1H-pyrrol-1-yl)aniline
OR32451-1G
OR32451-5G
OR32451-25G
APOSOR32451-1GEA
122
EUR
APOSOR32451-1G
APOSOR32451-5G
APOSOR32451-25G
2-(2,5-Dimethyl-1H-pyrrol-1-yl)phenylamine 95%
2-(2,5-Dimethyl-1H-pyrrol-1-yl)phenylamine
Formel:
C₁₂H₁₄N₂ Molecular Weight: 186.26 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01860278 CAS-Nummer: 2405-01-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...