1,2-Dihydronaphtho[2,1-b]furan-2-carboxylic acid 95%
Lieferant: Apollo Scientific
OR32453-1G
OR32453-5G
APOSOR32453-1GEA
270
EUR
APOSOR32453-1G
APOSOR32453-5G
1,2-Dihydronaphtho[2,1-b]furan-2-carboxylic acid 95%
1,2-Dihydronaphtho[2,1-b]furan-2-carboxylic acid
Formel:
C₁₃H₁₀O₃ Molecular Weight: 214.22 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03001250 CAS-Nummer: 24758-31-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...