5-Brompyrimidin-2-carbonitril
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Bromo-2-cyanopyrimidine, 5-bromo-pyrimidine-2-carbonitile
OR3247-1G
OR3247-5G
APOSOR3247-1GEA
21.9
EUR
APOSOR3247-1G
APOSOR3247-5G
5-Brompyrimidin-2-carbonitril
5-Brompyrimidin-2-carbonitril
Formel:
C₅H₂BrN₃ Molecular Weight: 183,99 g/mol Schmelzpunkt: 121…123 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02940446 CAS-Nummer: 38275-57-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...