1-(6,7-Dichloro-2,3-dihydro-1,4-benzodioxin-5-yl)-1-ethanone 95%
Lieferant: Apollo Scientific
OR32490-1G
OR32490-5G
APOSOR32490-1GEA
270
EUR
APOSOR32490-1G
APOSOR32490-5G
1-(6,7-Dichloro-2,3-dihydro-1,4-benzodioxin-5-yl)-1-ethanone 95%
1-(6,7-Dichloro-2,3-dihydro-1,4-benzodioxin-5-yl)-1-ethanone
Formel:
C₁₀H₈Cl₂O₃ Molecular Weight: 247.08 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03791175 CAS-Nummer: 166816-11-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...