DL-β-(3-Chlorphenyl)alaninol 95%

Lieferant: Apollo Scientific

Synonyme: 3-Amino-3-(3-chlorophenyl)propan-1-ol, 3-Amino-3-(3-chlorophenyl)-propan-1-ol

OR32505-1G
APOSOR32505-1GEA 270 EUR
APOSOR32505-1G
DL-β-(3-Chlorphenyl)alaninol 95%
DL-β-(3-Chlorphenyl)alaninol
Formel: C₉H₁₂ClNO
Molecular Weight: 185,65 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03412790
CAS-Nummer: 68208-25-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR