Methyl-2-amino-2-(4-chlorophenyl)acetate hydrochloride 97%
Lieferant: Apollo Scientific
Synonyme: Methyl 2-amino-2-(4-chlorophenyl)acetate hydrochloride
OR32590-25G OR32590-5G
APOSOR32590-25GEA 470 EUR
APOSOR32590-25G APOSOR32590-5G
Methyl-2-amino-2-(4-chlorophenyl)acetate hydrochloride 97%
Methyl-2-amino-2-(4-chlorophenyl)acetate hydrochloride
Formel: C₉H₁₁Cl₂NO₂
Molecular Weight: 236,1 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00196831
CAS-Nummer: 42718-19-4