2-Oxo-1,2-dihydro-3-pyridinecarbaldehyde 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Oxo-1,2-dihydropyridine-3-carbaldehyde

OR32598-1G OR32598-25G OR32598-5G
APOSOR32598-1GEA 29 EUR
APOSOR32598-1G APOSOR32598-25G APOSOR32598-5G
2-Oxo-1,2-dihydro-3-pyridinecarbaldehyde 95%
2-Oxo-1,2-dihydro-3-pyridinecarbaldehyde
Formel: C₆H₅NO₂
Molecular Weight: 123,11 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD07698588
CAS-Nummer: 36404-89-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR