5-Acetyl-6-methyl-2(1H)-pyridinone 95%
Lieferant: Apollo Scientific
Synonyme:
5-Acetyl-6-methyl-1,2-dihydropyridin-2-one, 5-Acetyl-6-methylpyridin-2(1H)-one
OR32644-1G
OR32644-5G
APOSOR32644-1GEA
179
EUR
APOSOR32644-1G
APOSOR32644-5G
5-Acetyl-6-methyl-2(1H)-pyridinone 95%
5-Acetyl-6-methyl-2(1H)-pyridinone
Formel:
C₈H₉NO₂ Molecular Weight: 151,17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01814812 CAS-Nummer: 5220-65-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...