1-Methyl-2-(4-methylpiperazino)-1H-indole-3-carbaldehyde 95%

Lieferant: Apollo Scientific

OR32658-1G OR32658-5G
APOSOR32658-1GEA 538 EUR
APOSOR32658-1G APOSOR32658-5G
1-Methyl-2-(4-methylpiperazino)-1H-indole-3-carbaldehyde 95%
1-Methyl-2-(4-methylpiperazino)-1H-indole-3-carbaldehyde
Formel: C₁₅H₁₉N₃O
Molecular Weight: 257,34 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05663789
CAS-Nummer: 886360-89-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR